missing translation for 'onlineSavingsMsg'
Få mere at vide

Thermo Scientific™ Denhardt's solution (50X)

Denhardt's solution (50X) is a mixture of blocking agents used in membrane-based hybridization protocols.

Tekniske data

Kemisk navn eller materiale Denhardt's solution
MDL nummer MFCD00717549
Koncentration 50X
Opløselighedsinformation Fully miscible.
Se flere specifikationer

Beskrivelse

Beskrivelse

Denhardt's solution is used to reduce non-specific binding of detection reagents to the Northern and Southern Blot membranes. Denhardt's Solution is a mixture of blocking agents used in membrane-based hybridization protocols.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description
• Formulated in 1966, Denhardt’s solution is a combination of high-molecular weight polymers used to prevent non-specific binding of nucleic acid probes to membranes and filters
• Denhardt’s solution contains 1% Ficoll (type 400), 1% polyvinylpyrrolidone, and 1% bovine serum albumin
Application
• Denhardt’s solution is used in Northern and Southern blotting to prevent non-specific binding of detection reagents to membranes
• It can be used along with gelatin-chrome alum in in situ hybridization assays to maintain chromosome integrity
• It has been used to facilitate quantitative DNA hybridization on nylon membranes
Tekniske data

Tekniske data

Denhardt's solution
MFCD00717549
50X
Fully miscible.
Videoer
Sikkerhed og håndtering

Sikkerhed og håndtering

missing translation for 'tsca' : Yes

Anbefalet opbevaring : Store at -20°C

SDS
Dokumenter

Dokumenter

Produktcertifikater

RUO – Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.