missing translation for 'onlineSavingsMsg'
Få mere at vide

Deltamethrin, TRC

Artikelnummer. 30004436
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004436 10 g
30008708 1 g
18060250 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30004436 Leverandør Toronto Research Chemicals Leverandørnr. TRCD23070010G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52918-63-5
Molekylær formel C22 H19 Br2 N O3
Molekylvægt (g/mol) 505.2
Synonym Cyclopropanecarboxylic acid, 3-(2,2-dibromoethenyl)-2,2-dimethyl-, cyano(3-phenoxyphenyl)methyl ester, [1R-[1α(S*),3α]]-, (S)-Cyano(3-phenoxyphenyl)methyl (1R,3R)-3-(2,2-dibromoethenyl)-2,2-dimethylcyclopropanecarboxylate, Alpha Max, AquaK'othrine, Battalion, Butox, Cislin, Crackdown, Decamethrin, Decamethrine, Decis, Decis 0.5ULV, Decis 1.5ULV, Decis 2.5EC, Decis 2.5ULV, Decis Expert, Decis Extra, Decis Flow, Decis Jet, Decis Super, Decis flow 2.5, Decistab, Decistab TB, Delta Gold, Delta Gold (insecticide), DeltaGard, Deltacid, Deltadust, Deltagran, Deltamethrin, Ezeetab, FMC 45498, Glossinex 200, IPO 8831, K-O Tab, K-Obiol, K-Obiol 2P, K-Othrin, K-Othrine, K-Othrine Flow 25, K-Othrine Polyzone, NRDC 161, New Musigie, Pendelta, Phagase 1, RU 22974, RUP 987, Scalibor, Spritex antiwasp powder, Suspend, Timbertreat DM 5, Ultratec, Vectocid, Zorcis
IUPAC navn [(S)-cyano-(3-phenoxyphenyl)methyl] (1R,3R)-3-(2,2-dibromoethenyl)-2,2-dimethylcyclopropane-1-carboxylate
SMIL CC1(C)[C@@H](C=C(Br)Br)[C@H]1C(=O)O[C@H](C#N)c2cccc(Oc3ccccc3)c2

Specifications

Kemisk navn eller materiale Deltamethrin
Renhedsgrad noter HPLC
Mængde 10 g
InChI formel InChI=1S/C22H19Br2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/t17-,18+,20-/m0/s1
Formel vægt 502.9732
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.