Beskrivelse
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Deltamethrin |
| Emballage | Glass Bottle |
| CAS | 52918-63-5 |
| Farve | Colorless |
| Smeltepunkt | 100°C to 102°C |
| Kogepunkt | Decomposes below the boiling point. |
| Synonym | (S)-α-Cyano-3-phenoxybenzyl (1R,3R)-3-(2,2-Dibromovinyl)-2,2-dimethylcyclopropanecarboxylate |
| Industry Type | Environmental Analysis |
| Molekylvægt (g/mol) | 505.2 |
| Mængde | 100 mg |
| Vis mere |