Få mere at vide
delta8,9-Dehydro Estrone 3-Sulfate Sodium (Stabilized with up to 40% Tromethamine), TRC
Artikelnummer. 30365378
Skift visning
Mængde:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30365378 1 mg
30357528 10 mg
30356216 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30365378 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2296451MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61612-83-7
Molekylær formel C18 H19 O5 S . Na
Molekylvægt (g/mol) 370.4
Synonym Estra-1,3,5(10),8-tetraen-17-one, 3-(sulfooxy)-, sodium salt (1:1), Estra-1,3,5(10),8-tetraen-17-one, 3-(sulfooxy)-, sodium salt (9CI), δ8,9-Dehydroestrone sodium sulfate
IUPAC navn sodium;[(13S,14S)-13-methyl-17-oxo-7,11,12,14,15,16-hexahydro-6H-cyclopenta[a]phenanthren-3-yl] sulfate
SMIL [Na+].C[C@]12CCC3=C(CCc4cc(OS(=O)(=O)[O-])ccc34)[C@@H]1CCC2=O
Tekniske data
Kemisk navn eller materiale delta 8,9-Dehydroestrone 3-Sulfate Sodium Salt
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C18H20O5S.Na/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;/h3,5,10,16H,2,4,6-9H2,1H3,(H,20,21,22);/q;+1/p-1/t16-,18-;/m0./s1
Formel vægt 370.0851
Procent renhed >95%