Få mere at vide
delta7-Stigmasterol, TRC
Artikelnummer. 30138138
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138138 50 mg
30010142 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138138 Leverandør Toronto Research Chemicals Leverandørnr. TRCS68676550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 481-19-6
Molekylær formel C29H46O
Molekylvægt (g/mol) 410.67
Synonym (3β)-Stigmasta-5,7,22-trien-3-ol, Stigmasta-5,7,22-trien-3β-ol, (24S)-Stigmasta-5,7,22-trien-3β-ol, 24(S)-24-Ethylcholesta-5,7,trans-22-trien-3β-ol, 7-Dehydrostigmasterol, Corbisterin, Corbisterol, δ5,7,22-Stigmastatrienol
IUPAC navn (3S,9S,10R,13R,14R,17R)-17-[(E,2R,5S)-5-ethyl-6-methylhept-3-en-2-yl]-10,13-dimethyl-2,3,4,9,11,12,14,15,16,17-decahydro-1H-cyclopenta[a]phenanthren-3-ol
SMIL O[C@H](C1)CC[C@@]2(C)C1=CC=C3[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)/C=C/[C@@H](CC)C(C)C
Tekniske data
Kemisk navn eller materiale delta7-Stigmasterol
Mængde 50 mg
InChI formel InChI=1S/C29H46O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h8-11,19-21,23,25-27,30H,7,12-18H2,1-6H3/b9-8+/t20-,21-,23+,25-,26+,27+,28+,29-/m1/s1
Formel vægt 410.3549