missing translation for 'onlineSavingsMsg'
Få mere at vide

delta6-Fulvestrant, TRC

Artikelnummer. 30137178
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137178 1 mg
30003324 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30137178 Leverandør Toronto Research Chemicals Leverandørnr. TRCF8625301MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 2170200-14-1
Molekylær formel C32H45F5O3S
Molekylvægt (g/mol) 604.75
Synonym 7-[9-[(4,4,5,5,5-Pentafluoropentyl)sulfinyl]nonyl]estra-1,3,5(10),6-tetraene-3,17β-diol, Fulvestrant Impurity E;
SMIL OC1=CC(C=C(CCCCCCCCCS(CCCC(F)(F)C(F)(F)F)=O)[C@]2([H])[C@]3([H])CC[C@@]4(C)[C@@]2([H])CC[C@@H]4O)=C3C=C1

Specifications

Kemisk navn eller materiale delta6-Fulvestrant
Mængde 1 mg
InChI formel InChI=1S/C32H45F5O3S/c1-30-17-15-26-25-12-11-24(38)21-23(25)20-22(29(26)27(30)13-14-28(30)39)10-7-5-3-2-4-6-8-18-41(40)19-9-16-31(33,34)32(35,36)37/h11-12,20-21,26-29,38-39H,2-10,13-19H2,1H3/t26-,27+,28+,29-,30+,41?/m1/s1
Formel vægt 604.301
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.