Få mere at vide
Delta2-Cephalexin, TRC
Artikelnummer. 30009146
Skift visning
Mængde:
5 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009146 100 mg
30003716 5 mg
30009019 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009146 Leverandør Toronto Research Chemicals Leverandørnr. TRCC256810100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 79750-46-2
Molekylær formel C16 H17 N3 O4 S
Molekylvægt (g/mol) 347.39
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic acid, 7-[[(2R)-aminophenylacetyl]amino]-3-methyl-8-oxo-, (6R,7R)- (9CI)
IUPAC navn (6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic acid
SMIL CC1=CS[C@@H]2[C@H](NC(=O)[C@H](N)c3ccccc3)C(=O)N2C1C(=O)O
Tekniske data
Kemisk navn eller materiale delta2-Cephalexin
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C16H17N3O4S/c1-8-7-24-15-11(14(21)19(15)12(8)16(22)23)18-13(20)10(17)9-5-3-2-4-6-9/h2-7,10-12,15H,17H2,1H3,(H,18,20)(H,22,23)/t10-,11-,12?,15-/m1/s1
Formel vægt 347.094
Procent renhed >95%