Få mere at vide
Delta2-Cefuroxime Axetil (Cefuroxime Axetil Delta-3 Isomer), TRC
Artikelnummer. 30012224
Skift visning
Mængde:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012224 25 mg
30004931 1 mg
17943805 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012224 Leverandør Toronto Research Chemicals Leverandørnr. TRCC24807025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 123458-61-7
Molekylær formel C20 H22 N4 O10 S
Molekylvægt (g/mol) 510.47
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic acid, 3-[[(aminocarbonyl)oxy]methyl]-7-[[(2Z)-2-(2-furanyl)-2-(methoxyimino)acetyl]amino]-8-oxo-, 1-(acetyloxy)ethyl ester, (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic acid, 3-[[(aminocarbonyl)oxy]methyl]-7-[[(2Z)-2-furanyl(methoxyimino)acetyl]amino]-8-oxo-, 1-(acetyloxy)ethyl ester, (6R,7R)- (9CI), 5-Thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic acid, 3-[[(aminocarbonyl)oxy]methyl]-7-[[2-furanyl(methoxyimino)acetyl]amino]-8-oxo-, 1-(acetyloxy)ethyl ester, [6R-[6α,7β(Z)]]-, δ2-Cefuroxime axetil
IUPAC navn 1-acetyloxyethyl (6R,7R)-3-(carbamoyloxymethyl)-7-[[(2Z)-2-(furan-2-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylate
SMIL CO\N=C(/C(=O)N[C@H]1[C@H]2SC=C(COC(=O)N)C(N2C1=O)C(=O)OC(C)OC(=O)C)\c3occc3
Tekniske data
Kemisk navn eller materiale Cefuroxime Axetil Delta-3-Isomers
Mængde 25 mg
InChI formel InChI=1S/C20H22N4O10S/c1-9(25)33-10(2)34-19(28)15-11(7-32-20(21)29)8-35-18-14(17(27)24(15)18)22-16(26)13(23-30-3)12-5-4-6-31-12/h4-6,8,10,14-15,18H,7H2,1-3H3,(H2,21,29)(H,22,26)/b23-13-/t10?,14-,15?,18-/m1/s1
Formel vægt 510.1057