Få mere at vide
Delta2-Ceftazidime (>80%), TRC
Artikelnummer. 30003862
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003862 10 mg
30009310 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30003862 Leverandør Toronto Research Chemicals Leverandørnr. TRCC24411010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1000980-60-8
Synonym Pyridinium, 1-[[(6R,7R)-7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-[(1-carboxy-1-methylethoxy)imino]acetyl]amino]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-en-3-yl]methyl]-, inner salt∣Ceftazidime Delta-3-Isomer
Tekniske data
Format Neat
Molekylvægt (g/mol) 546.58
Produkttype Impurity