Learn More
Chemsplash Delta 67 Coverall with Knit Wrist Cuff Type 5 and 6 Hood

Delta 67 Disposable Coverall is made from our soft comfortable 67 gsm microporous laminate fabric, meeting Cat III Type 5B/6B.
1250.00 DKK
Specifications
Product Type | Cleanroom Coveralls |
---|---|
Gender | Unisex |
Material | Microporous Composite Fabric |
Color | White |
Disposable | Single-use |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
30058331
![]() |
Chemsplash
2792.400.004 |
Small |
1250.00 DKK
Pack of 25 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
30058462
![]() |
Chemsplash
2792.400.005 |
Medium |
1250.00 DKK
Pack of 25 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
30058291
![]() |
Chemsplash
2792.400.006 |
Large |
1250.00 DKK
Pack of 25 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
30058455
![]() |
Chemsplash
2792.400.007 |
X-Large |
1250.00 DKK
Pack of 25 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
30058453
![]() |
Chemsplash
2792.400.008 |
2X-Large |
1250.00 DKK
Pack of 25 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
30058205
![]() |
Chemsplash
2792.400.009 |
3X-Large |
1250.00 DKK
Pack of 25 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
Description
Chemsplash Delta 67 Disposable Coverall
- 7GSM Microporous Laminate Fabric
- Elasticated 3 Piece Hood, Back and Ankles
- Low Linting Knitted Cuffs with Thumb Loops
- Two-Way Zip with Adhesive Zip Flap
- Reinforced Bound Seam at Crotch Area
- Non Linting and Anti-Static Fabric
- Silicone and Latex Free
- PFAS Free
- Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9
Description: The Chemsplash Delta 67 Disposable Coverall is made from soft, comfortable 67GSM microporous laminate fabric, meeting Cat III Type 5B/6B. It is compliant with EN1149-5:2008 (Antistatic) and EN1073-2 (Nuclear Particle). The coverall is ideal for various applications, particularly for prolonged wear, offering improved breathability while maintaining an effective barrier against hazards.
Specifications
Cleanroom Coveralls | |
Microporous Composite Fabric | |
Single-use | |
Elastic | |
Hooded | |
Ties | |
Non-sterile | |
Yes | |
Class 100-100,000 (ISO 5-8) |
Unisex | |
White | |
Elastic | |
Knit, Thumb Loop | |
3-panel | |
Stitched | |
Liquid | |
None |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.