missing translation for 'onlineSavingsMsg'
Få mere at vide

STEMCELL Technologies Delphinidin (Chloride)

Artikelnummer. 17120352
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17120352 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17120352 Leverandør STEMCELL Technologies Leverandørnr. 74232

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Inhibits EGFR and VEGFR

  • Delphinidin is an anthocyanidin found in pigmented fruits and vegetables with anti-inflammatory, anti-angiogenic, and anti-oxidant properties (Watson & Schönlau)
  • It has been reported to be a potent inhibitor of epidermal growth factor receptor (EGFR; Meiers et al.) and vascular endothelial growth factor receptor (VEGFR) (Lamy et al.) This product is supplied as the chloride salt of the molecule
  • Induces G2-M phase arrest in human prostate cancer PC3 cells, leading to apoptosis (Hafeez et al.)

Tekniske data

CAS 528-53-0
Synonym Delphinidol; Delphinidine; Ephdine
Mængde 1 mg
Molekylær formel C15H11ClO7
Renhed ≥97%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.