Få mere at vide
Delmadinone, TRC
Artikelnummer. 30366748
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366748 100 mg
30361667 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30366748 Leverandør Toronto Research Chemicals Leverandørnr. TRCD230655100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Delmadinone
CAS 15262-77-8
Molekylær formel C21 H25 Cl O3
Mængde 100 mg
Synonym Pregna-1,4,6-triene-3,20-dione, 6-chloro-17-hydroxy-, 6-Chloro-17-hydroxypregna-1,4,6-triene-3,20-dione, 6-Chloro-17α-hydroxypregna-1,4,6-triene-3,20-dione, Delmadinone, δ1-Chlormadinone
InChI formel InChI=1S/C21H25ClO3/c1-12(23)21(25)9-6-16-14-11-18(22)17-10-13(24)4-7-19(17,2)15(14)5-8-20(16,21)3/h4,7,10-11,14-16,25H,5-6,8-9H2,1-3H3/t14?,15-,16-,19+,20-,21-/m0/s1
SMIL CC(=O)[C@@]1(O)CC[C@H]2C3C=C(Cl)C4=CC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C
IUPAC navn (9S,10R,13S,14S,17R)-17-acetyl-6-chloro-17-hydroxy-10,13-dimethyl-9,11,12,14,15,16-hexahydro-8H-cyclopenta[a]phenanthren-3-one
Molekylvægt (g/mol) 360.87
Formel vægt 360.1492