Få mere at vide
Tocris Bioscience™ DEL 22379
GREENER_CHOICE
ERK dimerization inhibitor
Brand:  Tocris Bioscience™ 5774/10
Encompass
Produktkode 15757106
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
DEL 22379 is an ERK dimerization inhibitor (IC50 ∽ 0.5 μM); inhibits ERK dimerization without affecting phosphorylation or kinase activity. Suppresses growth of tumor cells expressing RAS-ERK oncogenes in vitro. Induces apoptosis and prevents tumor progression in vivo.
Tekniske data
Tekniske data
N-[2,3-Dihydro-3-[(5-methoxy-1H-indol-3-yl)methylene]-2-oxo-1H-indol-5-yl]-1-piperidinepropanamide
181223-80-3
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner