Få mere at vide
Tocris Bioscience™ DEL 22379
Beskrivelse
DEL 22379 is an ERK dimerization inhibitor (IC50 ∽ 0.5 μM); inhibits ERK dimerization without affecting phosphorylation or kinase activity. Suppresses growth of tumor cells expressing RAS-ERK oncogenes in vitro. Induces apoptosis and prevents tumor progression in vivo.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-[2,3-Dihydro-3-[(5-methoxy-1H-indol-3-yl)methylene]-2-oxo-1H-indol-5-yl]-1-piperidinepropanamide |
| CAS | 181223-80-3 |
| Mængde | 10 mg |
| Mål | ERK Inhibitors |
| Molekylær formel | C26H28N4O3 |
| Formel vægt | 444.53 |
| Renhed | >98% |