Få mere at vide
Tocris Bioscience™ DEL 22379
ERK dimerization inhibitor
Brand: Tocris Bioscience™ 5774/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
DEL 22379 is an ERK dimerization inhibitor (IC50 ∽ 0.5 μM); inhibits ERK dimerization without affecting phosphorylation or kinase activity. Suppresses growth of tumor cells expressing RAS-ERK oncogenes in vitro. Induces apoptosis and prevents tumor progression in vivo.Tekniske data
| N-[2,3-Dihydro-3-[(5-methoxy-1H-indol-3-yl)methylene]-2-oxo-1H-indol-5-yl]-1-piperidinepropanamide | |
| 181223-80-3 | |
| 50 mg |