Learn More
DEK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562975
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SEDSSDDEPL IKKLKKPPTD EELKETIKKL LASANLEEVT MKQICKKVYE NYPTYDLTER Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
DEK Polyclonal antibody specifically detects DEK in Human samples. It is validated for Immunohistochemistry.Specifications
DEK | |
Polyclonal | |
Unconjugated | |
DEK | |
1810019E15Rik; D13H6S231E; D6S231E; DEK; DEK oncogene (DNA binding); DEK proto-oncogene; protein DEK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7913 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P35659 | |
DEK | |
Recombinant protein corresponding to Human DEK | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only