missing translation for 'onlineSavingsMsg'
Få mere at vide

Dehydroepiandrosterone (Dhea) (16,16-D2, 97%), TRC

Artikelnummer. 30353215
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30353215 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30353215 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2295931MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Kemisk navn eller materiale Dehydroepiandrosterone 16,16-d2
CAS 67034-83-7
Molekylær formel C19 D2 H26 O2
Mængde 1 mg
Synonym Dehydro Epiandrosterone D2 (16,16-D2), Androst-5-en-17-one-16,16-d2, 3-hydroxy-, (3β)-, (3β)-3-Hydroxyandrost-5-en-17-one-16,16-d2, δ5-Androstene-3β-ol-17-one-16,16-d2, Dehydroepiandrosterone-16,16-D2, DHEA-d2, Dehydroepiandrosterone-d2, Prasterone-d2, Prasterone-16,16-d2
InChI formel InChI=1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h3,13-16,20H,4-11H2,1-2H3/t13-,14-,15-,16-,18-,19-/m0/s1/i6D2
SMIL [2H]C1([2H])C[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]2(C)C1=O
IUPAC navn (3S,8R,9S,10R,13S,14S)-16,16-dideuterio-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-17-one
Molekylvægt (g/mol) 290.4367
Formel vægt 290.2215
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.