Få mere at vide
Dehydroepiandrosterone-d6, TRC
Artikelnummer. 30359908
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30359908 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30359908 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2295881MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dehydroepiandrosterone (DHEA) (2,2,3,4,4,6-D6)
CAS 1261254-39-0
Molekylær formel C19 D6 H22 O2
Mængde 1 mg
Synonym Dehydro Epiandrosterone D6 (2,2,3,4,4,6-D6), Androst-5-en-17-one-2,2,3,4,4,6-d6, 3-hydroxy-, (3β)-, (3β)-3-Hydroxyandrost-5-en-17-one-2,2,3,4,4,6-d6, δ5-Androstene-3β-ol-17-one-2,2,3,4,4,6-d6, Dehydroepiandrosterone-2,2,3,4,4,6-D6, DHEA-d6, Dehydroepiandrosterone-d6, Prasterone-d6, Prasterone-2,2,3,4,4,6-d6
InChI formel InChI=1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h3,13-16,20H,4-11H2,1-2H3/t13?,14?,15?,16?,18-,19-/m0/s1/i3D,7D2,11D2,13D
SMIL [2H]C1=C2C([2H])([2H])C([2H])(O)C([2H])([2H])C[C@]2(C)C3CC[C@@]4(C)C(CCC4=O)C3C1
IUPAC navn (10R,13S)-2,2,3,4,4,6-hexadeuterio-3-hydroxy-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-one
Molekylvægt (g/mol) 294.46
Formel vægt 294.2466