Learn More
Dehydroacetic Acid Sodium Salt, Sigma-Aldrich™
Product Code. 15889780
Change view
Click to view available options
Quantity:
5 g
250 g
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15889780 250 g 250g
15879780 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15889780 Supplier Sigma Aldrich Supplier No. 8140850250
 Restricted Item
Forventet forsendelse: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4418-26-2
Specifications
Melting Point 280°C to 285°C
MDL Number MFCD00040583
Quantity 250 g
Synonym Dehydroacetic acid sodium salt, 3-(1-Hydroxyethylidene)-6-methyl-2H-pyran-2,4(3H)-dione sodium salt, Dehydroacetic acid sodium salt
Molecular Weight (g/mol) 190.13
Grade Research