Få mere at vide
Dehydro Olmesartan Medoxomil, TRC
Artikelnummer. 30003670
Skift visning
Mængde:
1 mg
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30003670 2.5 mg 2.5mg
30006416 25 mg -
30012687 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30003670 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2299652.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 879562-26-2
Molekylær formel C29H28N6O5
Molekylvægt (g/mol) 540.57
Synonym 1H-Imidazole-5-carboxylic acid, 4-(1-methylethenyl)-2-propyl-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl ester, 1H-Imidazole-5-carboxylic acid, 4-(1-methylethenyl)-2-propyl-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl ester, Olmesartan Medoxomil Imp C (EP)
IUPAC navn (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl 5-prop-1-en-2-yl-2-propyl-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylate
SMIL CCCc1nc(C(=C)C)c(C(=O)OCC2=C(C)OC(=O)O2)n1Cc3ccc(cc3)c4ccccc4c5nnn[nH]5
Tekniske data
Kemisk navn eller materiale Dehydro Olmesartan Medoxomil
Mængde 2.5 mg
InChI formel InChI=1S/C29H28N6O5/c1-5-8-24-30-25(17(2)3)26(28(36)38-16-23-18(4)39-29(37)40-23)35(24)15-19-11-13-20(14-12-19)21-9-6-7-10-22(21)27-31-33-34-32-27/h6-7,9-14H,2,5,8,15-16H2,1,3-4H3,(H,31,32,33,34)
Formel vægt 540.2121