Få mere at vide
Dehydro Norketamine-d4 (>90%), TRC
Artikelnummer. 30354277
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30354277 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30354277 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2299525MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dehydro Norketamine-d4
CAS 1246816-68-1
Molekylær formel C12H8D4ClNO
Mængde 5 mg
Synonym 6-Amino-6-(2-chlorophenyl-d4)-2-cyclohexen-1-one, 5,6-Dehydronorketamine-d4
InChI formel InChI=1S/C12H12ClNO/c13-10-6-2-1-5-9(10)12(14)8-4-3-7-11(12)15/h1-3,5-7H,4,8,14H2/i1D,2D,5D,6D
SMIL O=C1C(C2=C([2H])C([2H])=C([2H])C([2H])=C2Cl)(N)CCC=C1
Molekylvægt (g/mol) 225.75
Formel vægt 225.0858