Få mere at vide
Dehydro Indapamide-d3, TRC
Artikelnummer. 30355959
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30355959 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30355959 Leverandør Toronto Research Chemicals Leverandørnr. TRCD22966210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dehydro Indapamide-d3
CAS 1185057-48-0
Molekylær formel C16H11D3ClN3O3S
Mængde 10 mg
Synonym 3-(Aminosulfonyl)-4-chloro-N-[(2-methyl-d3)-1H-indol-1-yl]benzamide, 4-Chloro-3-sulfamoyl-N-[(2-methyl-d3)-1H-indol-1-yl]benzamide
InChI formel InChI=1S/C16H14ClN3O3S/c1-10-8-11-4-2-3-5-14(11)20(10)19-16(21)12-6-7-13(17)15(9-12)24(18,22)23/h2-9H,1H3,(H,19,21)(H2,18,22,23)/i1D3
SMIL O=C(C1=CC=C(Cl)C(S(N)(=O)=O)=C1)NN2C(C([2H])([2H])[2H])=CC3=CC=CC=C32
Molekylvægt (g/mol) 366.84