Få mere at vide
Dehydro Felodipine-d3, TRC
Artikelnummer. 30348814
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30348814 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30348814 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2296521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dehydro Felodipine-d3
CAS 1189916-52-6
Molekylær formel C18H14D3Cl2NO4
Mængde 1 mg
Synonym 4-(2,3-Dichlorophenyl)-2,6-dimethyl-3,5-pyridinedicarboxylic Acid Ethyl Methyl-d3 Ester, H 152/37-d3
InChI formel InChI=1S/C18H17Cl2NO4/c1-5-25-18(23)14-10(3)21-9(2)13(17(22)24-4)15(14)11-7-6-8-12(19)16(11)20/h6-8H,5H2,1-4H3/i4D3
SMIL O=C(OC([2H])([2H])[2H])C1=C(C2=CC=CC(Cl)=C2Cl)C(C(OCC)=O)=C(C)N=C1C
Molekylvægt (g/mol) 385.26