missing translation for 'onlineSavingsMsg'
Få mere at vide

Dehydro Aripiprazole, TRC

Artikelnummer. 17962715
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17962715 25 mg
17923625 5 mg
17954665 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17962715 Leverandør Toronto Research Chemicals Leverandørnr. TRCD22915525MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129722-25-4
Molekylær formel C23 H25 Cl2 N3 O2
Molekylvægt (g/mol) 446.37
Synonym 7-[4-[4-(2,3-Dichlorophenyl)piperazin-1-yl]butoxy]quinolin-2(1H)-one, Dehydroaripiprazole, Aripiprazole Related Compound G, Aripiprazole Imp E (EP), 2(1H)-Quinolinone, 7-[4-[4-(2,3-dichlorophenyl)-1-piperazinyl]butoxy]-, 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-2(1H)-quinolinone, DM 14857, OPC 14857, Dehydro Aripiprazole
IUPAC navn 7-[4-[4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy]-1H-quinolin-2-one
SMIL Clc1cccc(N2CCN(CCCCOc3ccc4C=CC(=O)Nc4c3)CC2)c1Cl

Specifications

Kemisk navn eller materiale Dehydro Aripiprazole
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C23H25Cl2N3O2/c24-19-4-3-5-21(23(19)25)28-13-11-27(12-14-28)10-1-2-15-30-18-8-6-17-7-9-22(29)26-20(17)16-18/h3-9,16H,1-2,10-15H2,(H,26,29)
Formel vægt 445.1324
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.