Få mere at vide
Dehydro Aripiprazole-d8 Hydrochloride, TRC
Artikelnummer. 30368665
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30368665 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30368665 Leverandør Toronto Research Chemicals Leverandørnr. TRCD22915210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dehydroaripiprazole piperazinyl-2,2,3,3,5,5,6,6-d8 hydrochloride
CAS 1215383-78-0
Renhedsgrad noter HPLC
Molekylær formel C23 2H8 H17 Cl2 N3 O2 . Cl H
Mængde 10 mg
Synonym 7-[4-[2,2,3,3,5,5,6,6-Octadeuterio-4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy]-1H-quinolin-2-one hydrochloride, Dehydro Aripiprazole-d8 hydrochloride, 7-[4-[4-(2,3-Dichlorophenyl) 2,2,3,3,5,5,6,6-d8-piperazin-1-yl]butoxy]quinolin-2(1H)-one hydrochloride, Dehydroaripiprazole-D8 hydrochloride (piperazinyl-2,2,3,3,5,5,6,6-D8), 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl-2,2,3,3,5,5,6,6-d8]butoxy]-2(1H)-quinolinone hydrochloride
InChI formel InChI=1S/C23H25Cl2N3O2.ClH/c24-19-4-3-5-21(23(19)25)28-13-11-27(12-14-28)10-1-2-15-30-18-8-6-17-7-9-22(29)26-20(17)16-18;/h3-9,16H,1-2,10-15H2,(H,26,29);1H/i11D2,12D2,13D2,14D2;
SMIL Cl.[2H]C1([2H])N(CCCCOc2ccc3C=CC(=O)Nc3c2)C([2H])([2H])C([2H])([2H])N(c4cccc(Cl)c4Cl)C1([2H])[2H]
IUPAC navn 7-[4-[2,2,3,3,5,5,6,6-octadeuterio-4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy]-1H-quinolin-2-one;hydrochloride
Molekylvægt (g/mol) 490.88
Formel vægt 489.1593
Procent renhed >95