Få mere at vide
Dehydro Amlodipine Besylate, TRC
Artikelnummer. 30135826
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135826 100 mg
30012410 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135826 Leverandør Toronto Research Chemicals Leverandørnr. TRCD228940100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2733700-89-3
Molekylær formel C20 23 Cl N2 O5 . C6 H6 O3 S
Molekylvægt (g/mol) 565.04
Synonym 3-ethyl 5-methyl 2-((2-aminoethoxy)methyl)-4-(2-chlorophenyl)-6-methylpyridine-3,5-dicarboxylate, benzenesulfonate (1:1)
IUPAC navn benzenesulfonic acid;O3-ethyl O5-methyl 2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methyl-pyridine-3,5-dicarboxylate
SMIL CCOC(=O)c1c(COCCN)nc(C)c(C(=O)OC)c1c2ccccc2Cl.OS(=O)(=O)c3ccccc3
Tekniske data
Kemisk navn eller materiale Dehydro Amlodipine Besylate
Mængde 100 mg
InChI formel InChI=1S/C20H23ClN2O5.C6H6O3S/c1-4-28-20(25)18-15(11-27-10-9-22)23-12(2)16(19(24)26-3)17(18)13-7-5-6-8-14(13)21;7-10(8,9)6-4-2-1-3-5-6/h5-8H,4,9-11,22H2,1-3H3;1-5H,(H,7,8,9)
Formel vægt 564.1333