Få mere at vide
Tocris Bioscience™ Deferoxamine mesylate
Iron chelator; also hypoxia mimetic
Brand: Tocris Bioscience™ 5764/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Deferoxamine mesylate is an iron chelator. Also hypoxia mimetic and inhibitor of ferroptosis. Has neuroprotective effects in animal models of stroke, traumatic brain injury and neurodegenerative diseases.Tekniske data
| N4-[5-[[4-[[5-(Acetylhydroxyamino)pentyl]amino-1,4-dioxobutyl]hydroxyamino]pentyl]-N1-(5-aminopentyl)-N1-hydroxybutanediamide methanesulfonate | |
| 138-14-7 | |
| 50 mg |