Få mere at vide
Deferiprone-d3, TRC
Artikelnummer. 30379327
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30379327 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30379327 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4740025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Deferiprone-d3
CAS 1346601-82-8
Molekylær formel C7H6D3NO2
Mængde 5 mg
Synonym 1,2-Dimethyl-3-hydroxypyrid-4-one-d3, 3-Hydroxy-1,2-dimethyl-4(1H)-pyridinone-d3, CP-20-d3, Kelfer-d3
InChI formel InChI=1S/C7H9NO2/c1-5-7(10)6(9)3-4-8(5)2/h3-4,10H,1-2H3/i2D3
SMIL O=C1C=CN(C([2H])([2H])[2H])C(C)=C1O
Molekylvægt (g/mol) 142.17
Formel vægt 142.0822