Få mere at vide
Deferasirox Ferrate(III) Tripotassium Complex Methanoate Hydrate, TRC
Artikelnummer. 30351262
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30351262 50 mg
1 options
Artikelnummer. 30351262 Leverandør Toronto Research Chemicals Leverandørnr. TRCD22867050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Deferasirox Ferrate(III) Tripotassium Complex Methanoate Hydrate
CAS 554445-58-8
Molekylær formel C44H36FeK3N6O12
Mængde 50 mg
Synonym (OC-6-22')-Bis[4-[3,5-bis[2-(hydroxy-kO)phenyl]-1H-1,2,4-triazol-1-yl-kN4]benzoato(3-)]ferrate(3-) Tripotassium Methanoate Hydrate
InChI formel InChI=1S/2C21H15N3O4.Fe/c2*25-17-7-3-1-5-15(17)19-22-20(16-6-2-4-8-18(16)26)24(23-19)14-11-9-13(10-12-14)21(27)28;/h2*1-12,25-26H,(H,27,28);/q;;+4/p-6
SMIL CO.CO.C1=C/C(=C/2\NN(/C(=C\3/C(=O)C=CC=C3)/N2)C4=CC=C(C=C4)C(=O)[O-])/C(=O)C=C1.C1=C/C(=C/2\NN(/C(=C\3/C(=O)C=CC=C3)/N2)C4=CC=C(C=C4)C(=O)[O-])/C(=O)C=C1.O.O.[K+].[K+].[K+].[Fe]
IUPAC navn 4-[3,5-bis(2-oxidophenyl)-1,2,4-triazol-1-yl]benzoate;iron(4+)
Molekylvægt (g/mol) 1013.93