Få mere at vide
Novus Biologicals™ DEFB107A Recombinant Protein Antigen
Artikelnummer. 18264543 Shop alle Bio Techne produkter
Skift visning
Protein længde:
EFELDRICGYGTARCRKKCRSQEYRIGRCPNTYACCLRKWDESLLNRT
QARTAIHRALISKRMEGHCEAECLTFEVKIGGCRAELAP
Pakningsstørrelse:
0.1ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Protein længde unitSize
18264543 EFELDRICGYGTARCRKKCRSQEYRIGRCPNTYACCLRKWDESLLNRT -
18277151 QARTAIHRALISKRMEGHCEAECLTFEVKIGGCRAELAP 0.1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18264543 Leverandør Novus Biologicals™ Leverandørnr. NBP259792PEP
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human DEFB104A The DEFB104A Recombinant Protein Antigen is derived from E. coli. The DEFB104A Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY
Tekniske data
Protein længde EFELDRICGYGTARCRKKCRSQEYRIGRCPNTYACCLRKWDESLLNRT
Oprensningsmetode >80% by SDS-PAGE and Coomassie blue staining
Fælles navn DEFB104A Recombinant Protein Antigen
Indhold og opbevaring Store at −20°C. Avoid freeze-thaw cycles.
Formulering PBS and 1M Urea, pH 7.4
Til brug med (applikation) AC
Gene Alias DEFB104B, DEFB-4, defensin
Gen symbol DEFB104A
Etikettype Unlabeled
Produkttype Recombinant Protein Antigen
Mængde 0.1 mL
Regulatorisk status RUO
Kilde E.coli
Specifik reaktivitet Human
Vis mere Vis mindre
For Research Use Only.