Få mere at vide
Deethylcyanazine acid, TRC
Artikelnummer. 17993155
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17993155 25 mg
17991235 50 mg
17902325 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17993155 Leverandør Toronto Research Chemicals Leverandørnr. TRCD95350025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 36749-35-6
Molekylær formel C7 H10 Cl N5 O2
Molekylvægt (g/mol) 231.64
Synonym Cyanazine acid-desethyl, N-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-2-methylalanine, 2-Chloro-4-(1-carbonyl-1-methylethylamino)-6-amino-1,3,5-triazine, N-(4-amino-6-chloro-1,3,5-triazin-2-yl)-2-methyl-Alanine
IUPAC navn 2-[(4-amino-6-chloro-1,3,5-triazin-2-yl)amino]-2-methyl-propanoic acid
SMIL CC(C)(Nc1nc(N)nc(Cl)n1)C(=O)O
Tekniske data
Kemisk navn eller materiale Deethylcyanazine acid
Mængde 25 mg
InChI formel InChI=1S/C7H10ClN5O2/c1-7(2,3(14)15)13-6-11-4(8)10-5(9)12-6/h1-2H3,(H,14,15)(H3,9,10,11,12,13)
Formel vægt 231.0523