Få mere at vide
DEET-d10, TRC
Artikelnummer. 30366512
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30366512 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366512 Leverandør Toronto Research Chemicals Leverandørnr. TRCD22850210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Deet-d10
CAS 1215576-01-4
Renhedsgrad noter HPLC
Molekylær formel C12H7D10NO
Mængde 10 mg
Synonym DEET D10 (diethyl D10), 3-Methyl-N,N-bis(1,1,2,2,2-pentadeuterioethyl)benzamide, Diethyltoluamide-d10
InChI formel InChI=1S/C12H17NO/c1-4-13(5-2)12(14)11-8-6-7-10(3)9-11/h6-9H,4-5H2,1-3H3/i1D3,2D3,4D2,5D2
SMIL [2H]C([2H])([2H])C([2H])([2H])N(C(=O)c1cccc(C)c1)C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn 3-methyl-N,N-bis(1,1,2,2,2-pentadeuterioethyl)benzamide
Molekylvægt (g/mol) 201.33
Procent renhed >95