Learn More
Thermo Scientific™ MaxQ™ 8000 Shakers Dedicated Platforms with Clamps
Maximize vessel capacity and simplicity when working with a single vessel size such as a flask using Thermo Scientific™ MaxQ™ 8000 Shakers Dedicated Platforms with Clamps.
Brand:  Thermo Scientific™ 238073
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10013982
  • 10220.00 DKK / Each
Estimated Shipment
16-02-2021
Add to basket
Description
Description
  • Clamps come installed to provide maximum capacity
Specifications
Specifications
12
45.7cm
Shaker Platform with Clamps
75cm
2L Erlenmeyer Flask
MaxQ™ 8000 Shaker