Få mere at vide
Decoquinate 100 μg/mL in Acetonitrile/DMF, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Decoquinate |
| CAS | 18507-89-6 |
| Analyt- eller komponentnavne | Decoquinate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C24 H35 N O5 |
| Koncentration | 100 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | 3-Quinolinecarboxylic acid, 6-(decyloxy)-7-ethoxy-4-hydroxy-, ethyl ester, Deccox, Decoquinate, Decoxy, Ethyl 4-hydroxy-6-(decyloxy)-7-ethoxyquinoline-3-carboxylate, Ethyl 6-decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylate, Ethyl 6-n-decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylate, HC 1528, M and B 15497, NSC 339057 |
| InChI formel | InChI=1S/C24H35NO5/c1-4-7-8-9-10-11-12-13-14-30-21-15-18-20(16-22(21)28-5-2)25-17-19(23(18)26)24(27)29-6-3/h15-17H,4-14H2,1-3H3,(H,25,26) |
| SMIL | CCCCCCCCCCOc1cc2c(O)c(cnc2cc1OCC)C(=O)OCC |
| Vis mere |