missing translation for 'onlineSavingsMsg'
Learn More

Groesfjeld Diagnostics deconex™ 11 Universal

Suitable for soaking baths and ultrasonic cleaning equipment. Groesfjeld Diagnostics deconex™ 11 Universal is a mildly alkaline concentrate with activated chlorine for decontaminating lab glassware. It can be universally used in different sectors and is an excellent substitute for chromic acid.

Brand:  Groesfjeld Diagnostics 20105

Additional Details : Weight : 5.50000kg Transport : UN number : 3266 Chem class : 8 Pack group : II


 View more versions of this product

Product Code. 11496454

Quantity Price
1 2245.00 DKK / 5 L
6 2028.58 DKK / 5 L
Estimated Shipment
31-05-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

deconex™ 11 Universal exhibits a wide cleaning spectrum due to its composition. This allows even stubborn contaminants to be removed, such as baked-on fats, oils, waxes, dried blood, proteins, residues of culture media, pen markings, pigments, traces of organic and inorganic materials, and silicones.

  • The surfactants used result in excellent wetting of glassware and can be rinsed without residue
  • The combination of surfactants with the alkalis and activated chlorine removes contaminants and prevents them from being re-deposited on the glassware
  • The easily biodegradable product is pH-buffered and intended for use over a period of several days
  • pH (1% solution in demineralized water): approx. 11.8
  • Density (in concentrate): 1.10g/ml
Specifications

Specifications

Suitable for borosilicate, Duran and quartz glass; porcelain; stainless and chrome nickel steel; polyethylene, polypropylene, Teflon, silicone and natural rubber. Not suitable for aluminium, zinc or tin.
White emulsion
5L
Alkalis, surfactants, inorganic chlorine carrier
Concentrated Liquid
Cleaning and decontamination
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • deconex 11 UNIVERSAL

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 B
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.

Supplemental information
  • MIXTURE LIST-Contains : Tetracalcium phosphate, potassium hydroxide, sodium hypochlorite

SDS
Documents