missing translation for 'onlineSavingsMsg'
Få mere at vide

Dechloro Chlonixin, TRC

Artikelnummer. 30350287
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30350287 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30350287 Leverandør Toronto Research Chemicals Leverandørnr. TRCD226745250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Dechloro Chlonixin
CAS 17782-05-7
Molekylær formel C13H12N2O2
Mængde 250 mg
Synonym 2-[(2-Methylphenyl)amino]-3-pyridinecarboxylic Acid, 2-o-Toluidino-nicotinic Acid, 2-(2-Methylanilino)nicotinic Acid
InChI formel InChI=1S/C13H12N2O2/c1-9-5-2-3-7-11(9)15-12-10(13(16)17)6-4-8-14-12/h2-8H,1H3,(H,14,15)(H,16,17)
SMIL Cc1ccccc1Nc2ncccc2C(=O)O
IUPAC navn 2-(2-methylanilino)pyridine-3-carboxylic acid
Molekylvægt (g/mol) 228.25
Formel vægt 228.0899
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.