Få mere at vide
Dechlorane A, TRC
Artikelnummer. 17958394
Skift visning
Mængde:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17958394 1 mg
17928674 25 mg
17948304 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17958394 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2262151MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13560-89-9
Molekylær formel C18 H12 Cl12
Molekylvægt (g/mol) 653.72
Synonym 1,4:7,10-Dimethanodibenzo[a,e]cyclooctene, 1,2,3,4,7,8,9,10,13,13,14,14-dodecachloro-1,4,4a,5,6,6a,7,10,10a,11,12,12a-dodecahydro-, 1,2,3,4,7,8,9,10,13,13,14,14-Dodecachloro-1,4,4a,5,6,6a,7,10,10a,11,12,12a-dodecahydro-1,4:7,10-dimethanodibenzo[a,e]cyclooctene, 1,2,3,4,7,8,9,10,13,13,14,14-Dodecachloro-1,4,4a,5,6,6a,7,10,10a,11,12,12a-dodecahydro-1,4:7,10-dimethanodibenzo[a,e]cyclooctane, Bis(hexachlorocyclopentadieno)cyclooctane, Dechloran A, Dechlorane Plus, Dechlorane Plus 1000, Dechlorane Plus 25, Dechlorane Plus 2520, Dechlorane Plus 35, Dechlorane Plus 515, Dodecachlorodimethanodibenzocyclooctane, Dodecachlorododecahydrodimethanodibenzocyclooctane, Dodecachlorododecahydrodimethanodibenzocyclooctene
IUPAC navn 1,6,7,8,9,14,15,16,17,17,18,18-dodecachloropentacyclo[12.2.1.16,9.02,13.05,10]octadeca-7,15-diene
SMIL ClC1=C(Cl)[C@@]2(Cl)C3CCC4C(CCC3[C@]1(Cl)C2(Cl)Cl)[C@@]5(Cl)C(=C(Cl)[C@]4(Cl)C5(Cl)Cl)Cl
Tekniske data
Kemisk navn eller materiale Dechlorane A
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C18H12Cl12/c19-9-10(20)15(25)7-3-4-8-6(2-1-5(7)13(9,23)17(15,27)28)14(24)11(21)12(22)16(8,26)18(14,29)30/h5-8H,1-4H2/t5?,6?,7?,8?,13-,14-,15+,16+
Formel vægt 647.7201
Procent renhed >95%