Få mere at vide
Decarboxylated S-Adenosylmethionine Sulfate Salt, TRC
Artikelnummer. 30007189
Skift visning
Mængde:
1 mg
2.5 mg
10 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30007189 10 mg -
30004369 2.5 mg 2.5mg
17968874 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30007189 Leverandør Toronto Research Chemicals Leverandørnr. TRCD22200010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 67380-81-8
Molekylær formel C14H23N6O3S • H2SO4 • HSO4-
Molekylvægt (g/mol) 355.4498079706
Synonym 5'-[(3-aminopropyl)methylsulfonio]-5'-deoxy-adenosine Sulfate (Salt) Sulfate (Salt) (1:1:1), (-)-S-Adenosyl-(5')-3-methylthiopropylamine Sulfate Salt, Decarboxylated AdoMet Sulfate Salt, S-Adenosyl-3-methylthiopropylamine Sulfate Salt, S-Adenosyl-L-methionamine Sulfate Salt, S-Adenosylmethionamine Sulfate Salt, S-Methyl-S-adenosylhomocysteamine Sulfate Salt, S-Methyladenosylhomocysteamine Sulfate Salt, dAdoMet Sulfate Salt;
IUPAC navn 3-aminopropyl-[[(2S,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl]-methylsulfanium;hydrogen sulfate;sulfuric acid
SMIL O=S(=O)(O)[O-].NC1=NC=NC2=C1N=CN2[C@H]3[C@H](O)[C@H](O)[C@@H](C[S+](C)CCCN)O3.[H2SO4]
Tekniske data
Kemisk navn eller materiale Decarboxylated S-Adenosylmethionine Sulfate Salt
Mængde 10 mg
InChI formel InChI=1S/C14H23N6O3S.2H2O4S/c1-24(4-2-3-15)5-8-10(21)11(22)14(23-8)20-7-19-9-12(16)17-6-18-13(9)20;2*1-5(2,3)4/h6-8,10-11,14,21-22H,2-5,15H2,1H3,(H2,16,17,18);2*(H2,1,2,3,4)/q+1;;/p-1/t8-,10-,11-,14-,24?;;/m1../s1