Få mere at vide
Decanoic-d19 Acid, TRC
Artikelnummer. 30349818
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30349818 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30349818 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28813050MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Decanoic acid-d19
CAS 88170-22-3
Molekylær formel C10 D19 H O2
Mængde 50 mg
Synonym Decanoic-d19 acid (9CI, ACI), d19-Decanoic acid, Nonadecadeuteriodecanoic acid, Decanoic acid D19
InChI formel InChI=1S/C10H20O2/c1-2-3-4-5-6-7-8-9-10(11)12/h2-9H2,1H3,(H,11,12)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O
IUPAC navn 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecadeuteriodecanoic acid
Molekylvægt (g/mol) 191.38
Formel vægt 191.2656