Få mere at vide
Decanoic Acid-1,2-13C2, TRC
Artikelnummer. 30363509
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30363509 5 mg
30360368 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30363509 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2125275MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 287111-30-2
Molekylær formel C8 13C2 H20 O2
Molekylvægt (g/mol) 174.25
Synonym 1-Decanoic Acid-13C2, 1-Nonanecarboxylic Acid-13C2, Capric Acid-13C2, Caprinic Acid-13C2, Caprynic Acid-13C2, Decoic Acid-13C2, Decylic Acid-13C2, Emery 659-13C2, Lunac 10-95-13C2, Lunac 10-98-13C2, Lunac 10-98E-13C2, NAA 102-13C2, NSC 5025-13C2, Prifac 2906-13C2, Prifac 296-13C2, n-Capric Acid-13C2, n-Decanoic Acid-13C2, n-Decoic Acid-13C2, n-Decylic Acid-13C2
IUPAC navn (1,2-^{13}C_{2})decanoic acid
SMIL CCCCCCCC[13CH2][13C](=O)O
Tekniske data
Kemisk navn eller materiale Decanoic Acid-1,2-13C2
Mængde 5 mg
InChI formel InChI=1S/C10H20O2/c1-2-3-4-5-6-7-8-9-10(11)12/h2-9H2,1H3,(H,11,12)/i9+1,10+1
Formel vægt 174.153