Få mere at vide
Decanoic-9,9,10,10,10-d5 Acid, TRC
Artikelnummer. 30366269
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366269 10 mg
30363522 5 mg
30352341 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366269 Leverandør Toronto Research Chemicals Leverandørnr. TRCD21252910MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1219803-00-5
Molekylær formel C10 D5 H15 O2
Molekylvægt (g/mol) 177.3
Synonym 9,9,10,10,10-Pentadeuteriodecanoic acid, Decanoic acid-9,9,10,10,10-d5
IUPAC navn 9,9,10,10,10-pentadeuteriodecanoic acid
SMIL [2H]C([2H])([2H])C([2H])([2H])CCCCCCCC(=O)O
Tekniske data
Kemisk navn eller materiale Decanoic-9,9,10,10,10-d5 Acid
Mængde 10 mg
InChI formel InChI=1S/C10H20O2/c1-2-3-4-5-6-7-8-9-10(11)12/h2-9H2,1H3,(H,11,12)/i1D3,2D2
Formel vægt 177.1777