Learn More
Decane, ≥99%, Reagent Grade, Honeywell™
Catalog Number p-7120367
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
124-18-5
C10H22
142.286
MFCD00008954
DIOQZVSQGTUSAI-UHFFFAOYSA-N
n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
15600
CHEBI:41808
decane
CCCCCCCCCC
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
124-18-5
C10H22
142.286
MFCD00008954
DIOQZVSQGTUSAI-UHFFFAOYSA-N
n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
15600
CHEBI:41808
decane
CCCCCCCCCC
Reagent Grade,≥99%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Boiling Range 174° - 180°C
Melting Point -27°C
Color Colorless
Density 0.73g/mL at 25°C
Explosion Limit 0.026
Flash Point 46°C (114.8°F)
Ignition Point 410°F
Odor Odorless
Packaging Glass bottle
Linear Formula CH3(CH2)8CH3
Quantity 100 mL
UN Number UN2247
Beilstein 1696981
Vapor Density 4.9 (vs air)
Formula Weight 142.28g/mol
Vapor Pressure 3.77mmHg (37.7°C), 1mmHg (16.5°C)
Percent Purity ≥99%
Chemical Name or Material Decane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Decane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.