missing translation for 'onlineSavingsMsg'
Få mere at vide

Decamethyl-1,9-Pentasiloxanediol, TRC

Artikelnummer. 30366547
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366547 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30366547 Leverandør Toronto Research Chemicals Leverandørnr. TRCD21246010MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Decamethyl-1,9-Pentasiloxanediol
CAS 7445-36-5
Molekylær formel C10H32O6Si5
Mængde 10 mg
Synonym 1,1,3,3,5,5,7,7,9,9-Decamethyl-1,9-pentasiloxanediol, 1,9-Dihydroxydecamethylpentasiloxane, Decamethylpentasiloxane-1,9-diol, Decamethylpentasiloxanediol
InChI formel InChI=1S/C10H32O6Si5/c1-17(2,11)13-19(5,6)15-21(9,10)16-20(7,8)14-18(3,4)12/h11-12H,1-10H3
SMIL C[Si](C)(O)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O
IUPAC navn bis[[[hydroxy(dimethyl)silyl]oxy-dimethylsilyl]oxy]-dimethylsilane
Molekylvægt (g/mol) 388.78
Produkttitel
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.