Få mere at vide
Decalin-d18 (cis/trans mixture), CDN
Artikelnummer. 30326092
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326092 5 g
30322226 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326092 Leverandør CDN Leverandørnr. CDND08855G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Decalin-d18
CAS 28788-42-3
Molekylær formel C10 D18
Mængde 5 g
Synonym Naphthalene-1,2,3,4,5,6,7,8-d8, decahydro-d10- (ACI), Decahydro-d10naphthalene-1,2,3,4,5,6,7,8-d8 (ACI), Naphthalene-d8, decahydro-d10- (8 CI, 9 CI), 1,1,2,2,3,3,4,4,4 A,5,5,6,6,7,7,8,8,8 A-Octadecadeuterionaphthalene, Decahydronaphthalene-d18, Decahydronaphthalene-D18, Decalin-d18
InChI formel InChI=1 S/C10H18/c1-2-6-10-8-4-3-7-9(10)5-1/h9-10 H,1-8H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9 D,10 D
SMIL [2 H]C1([2 H])C([2 H])([2 H])C([2 H])([2 H])C2([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C2([2 H])C1([2 H])[2 H]
IUPAC navn 1,1,2,2,3,3,4,4,4 A,5,5,6,6,7,7,8,8,8 A-octadecadeuterionaphthalene
Molekylvægt (g/mol) 156.3608
Formel vægt 156.2538 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity