missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
DEC1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-1800PE
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
DEC1 Polyclonal antibody specifically detects DEC1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Tekniske data
| DEC1. | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| PE | |
| basic helix-loop-helix domain containing, class B, 2, basic helix-loop-helix family, member e40, bHLHb2, bHLHe40, Class B basic helix-loop-helix protein 2, class E basic helix-loop-helix protein 40, DEC1HLHB2, differentially expressed in chondrocytes 1, Differentially expressed in chondrocytes protein 1, differentiated embryo chondrocyte expressed gene 1, Enhancer-of-split and hairy-related protein 2, SHARP2, SHARP-2, Stimulated by retinoic acid gene 13 protein, STRA13FLJ99214, Stra14 | |
| Synthetic peptide made to a C-terminal region of human DEC1 (between amino acids 350-412) [UniProt O14503] | |
| 0.1 mL | |
| HIF Target Genes, Hypoxia | |
| 8553 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion