Få mere at vide
Debromo Chloro Nicergoline, TRC
Artikelnummer. 30368215
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30368215 5 mg
30362568 10 mg
30360450 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30368215 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2088005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 38536-28-6
Molekylær formel C24 H26 Cl N3 O3
Molekylvægt (g/mol) 439.93
Synonym [(6aR,9R,10aS)-10a-Methoxy-4,7-dimethyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinolin-9-yl]methyl 5-Chloropyridine-3-carboxylate, Nicergoline Imp. A (EP), Chloronicergoline, Ergoline-8-methanol, 10-methoxy-1,6-dimethyl-, 8-[5-chloro-3-pyridinecarboxylate], Ergoline-8-methanol, 10-methoxy-1,6-dimethyl-, 5-chloro-3-pyridinecarboxylate (ester), (8β)- (9CI), [(8β)-10-Methoxy-1,6-dimethylergolin-8-yl]methyl 5-chloro-3-pyridinecarboxylate
IUPAC navn [(6aR,9R,10aS)-10a-methoxy-4,7-dimethyl-6a,8,9,10-tetrahydro-6H-indolo[4,3-fg]quinoline-9-yl]methyl 5-chloropyridine-3-carboxylate
SMIL CO[C@]12C[C@@H](COC(=O)c3cncc(Cl)c3)CN(C)[C@@H]1Cc4cn(C)c5cccc2c45
Tekniske data
Kemisk navn eller materiale Chloronicergoline
Mængde 5 mg
InChI formel InChI=1S/C24H26ClN3O3/c1-27-13-17-8-21-24(30-3,19-5-4-6-20(27)22(17)19)9-15(12-28(21)2)14-31-23(29)16-7-18(25)11-26-10-16/h4-7,10-11,13,15,21H,8-9,12,14H2,1-3H3/t15-,21-,24+/m1/s1
Formel vægt 439.1663