Få mere at vide
Debrisoquin Sulfate, Mikromol™
Artikelnummer. 17942346
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
17942346 100 mg
1 options
Artikelnummer. 17942346 Leverandør Mikromol Leverandørnr. MM0594.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Debrisoquin Sulfate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Debrisoquine Sulfate
CAS 581-88-4
Analyt- eller komponentnavne Debrisoquine Sulfate
Forsendelsestilstand Room Temperature
Molekylær formel 2 C10 H13 N3 . H2 O4 S
Certificeringer/compliance ISO 17025
Synonym 2(1H)-Isoquinolinecarboximidamide, 3,4-dihydro-, sulfate (2:1), 2(1H)-Isoquinolinecarboxamidine, 3,4-dihydro-, sulfate (2:1) (8CI), 1,2,3,4-Tetrahydro-isoquinoline-2-carboxamidine sulfate, Debrisoquin hemisulfate, Debrisoquin sulfate, Debrisoquine hemisulfate, Debrisoquine sulfate, Declinax, Isocaramidine sulfate, Ro 5-3307/1, Debrisoquin Sulphate
InChI formel InChI=1S/2C10H13N3.H2O4S/c2*11-10(12)13-6-5-8-3-1-2-4-9(8)7-13;1-5(2,3)4/h2*1-4H,5-7H2,(H3,11,12);(H2,1,2,3,4)
SMIL NC(=N)N1CCc2ccccc2C1.NC(=N)N3CCc4ccccc4C3.OS(=O)(=O)O
IUPAC navn 3,4-dihydro-1H-isoquinoline-2-carboximidamide;sulfuric acid
Molekylvægt (g/mol) 448.54
Mængde 100 mg
Formel vægt 448.1893 g/mol