Få mere at vide
Debrisoquin Sulfate, LoGiCal
Artikelnummer. 30462096
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30462096 10 mg
1 options
Artikelnummer. 30462096 Leverandør LoGiCal Leverandørnr. LGCFOR0594.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Debrisoquine Sulfate
CAS 581-88-4
Analyt- eller komponentnavne Debrisoquine Sulfate
Forsendelsestilstand Room Temperature
Molekylær formel 2 C10 H13 N3 . H2 O4 S
Certificeringer/compliance ISO 9001
Synonym 2(1H)-Isoquinolinecarboximidamide, 3,4-dihydro-, sulfate (2:1), 2(1H)-Isoquinolinecarboxamidine, 3,4-dihydro-, sulfate (2:1) (8CI), 1,2,3,4-Tetrahydro-isoquinoline-2-carboxamidine sulfate, Debrisoquin hemisulfate, Debrisoquin sulfate, Debrisoquine hemisulfate, Debrisoquine sulfate, Declinax, Isocaramidine sulfate, Ro 5-3307/1, Debrisoquin Sulphate
InChI formel InChI=1S/2C10H13N3.H2O4S/c2*11-10(12)13-6-5-8-3-1-2-4-9(8)7-13;1-5(2,3)4/h2*1-4H,5-7H2,(H3,11,12);(H2,1,2,3,4)
Løsningstype Neat
SMIL NC(=N)N1CCc2ccccc2C1.NC(=N)N3CCc4ccccc4C3.OS(=O)(=O)O
IUPAC navn 3,4-dihydro-1H-isoquinoline-2-carboximidamide;sulfuric acid
Industry Type Clinical Testing
Molekylvægt (g/mol) 448.54
Mængde 10 mg
Formel vægt 448.1893 g/mol