Få mere at vide
DEAE-Cellulose, (approx. 70% moisture content), TRC
Artikelnummer. 30003488
Skift visning
Mængde:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003488 50 g
30008527 10 g
30003598 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30003488 Leverandør Toronto Research Chemicals Leverandørnr. TRCD19905050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 9013-34-7
Molekylær formel C18H38N2O6
Molekylvægt (g/mol) 378.5
Synonym Cellulose-DEAE, Diethylaminoethyl-cellulose
IUPAC navn (2R,3R,4S,5S,6R)-3-[2-(diethylamino)ethoxy]-6-[2-(diethylamino)ethoxymethyl]oxane-2,4,5-triol
SMIL CCN(CC)CCOC[C@@H]1[C@H]([C@@H]([C@H]([C@@H](O1)O)OCCN(CC)CC)O)O
Tekniske data
Kemisk navn eller materiale DEAE-Cellulose
Mængde 50 g
InChI formel InChI=1S/C18H38N2O6/c1-5-19(6-2)9-11-24-13-14-15(21)16(22)17(18(23)26-14)25-12-10-20(7-3)8-4/h14-18,21-23H,5-13H2,1-4H3/t14-,15-,16+,17-,18-/m1/s1