Få mere at vide
Deacetylcephalothin Sodium Salt, TRC
Artikelnummer. 30366053
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366053 100 mg
30363197 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30366053 Leverandør Toronto Research Chemicals Leverandørnr. TRCD198210100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5547-29-5
Molekylær formel C14H13N2NaO5S2
Molekylvægt (g/mol) 376.38
Synonym (6R,7R)-3-(Hydroxymethyl)-8-oxo-7-[[2-(2-thienyl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Sodium Salt, (6R,7R)- 3-(Hydroxymethyl)-8-oxo-7-[(2-thienylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Sodium Salt, (6R-trans)-3-(Hydroxymethyl)-8-oxo-7-[(2-thienylacetyl)amino]-5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Sodium Salt, , 3-(Hydroxymethyl)-8-oxo-7-[2-(2-thienyl)acetamido]-5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Sodium Salt, (6R,7R)-3-Hydroxymethyl-7-(thien-2-ylacetamido)ceph-3-em-4-carboxylic Acid Sodium Salt, 3-Hydroxymethyl-7-(2-thienylacetamide)-3-cephem-4-carboxylic Acid Sodium Salt, Deacetylcephalothin Sodium Salt, Deacetylcephalotin Sodium Salt, Desacetylcephalothin Sodium Salt
IUPAC navn sodium;(6R,7R)-3-(hydroxymethyl)-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMIL [Na+].OCC1=C(N2[C@H](SC1)[C@H](NC(=O)Cc3cccs3)C2=O)C(=O)[O-]
Tekniske data
Kemisk navn eller materiale Deacetylcephalothin Sodium Salt
Mængde 100 mg
InChI formel InChI=1S/C14H14N2O5S2.Na/c17-5-7-6-23-13-10(12(19)16(13)11(7)14(20)21)15-9(18)4-8-2-1-3-22-8;/h1-3,10,13,17H,4-6H2,(H,15,18)(H,20,21);/q;+1/p-1/t10-,13-;/m1./s1
Formel vægt 376.0164